Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-4 Mouse, anti-Mouse, R718, Clone: 21H12, BD OptiBuild™

Catalog No. BDB752180 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752180 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752180 Supplier BD Biosciences Supplier No. 752180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 21H12 monoclonal antibody specifically binds to TIM-4. TIM-4 is encoded by Timd4 (T cell immunoglobulin and mucin domain containing 4). TIM-4 is also known as Spleen, mucin-containing, knockout of lymphotoxin protein (SMUCKLER). TIM-4 is a single-pass type I membrane transmembrane glycoprotein belonging to the TIM family of the immunoglobulin superfamily. TIM-4 is expressed by macrophages and at low levels by dendritic cells. TIM-4 is a phosphatidylserine receptor that enhances the phagocytosis of apoptotic cells. It can also serve as a receptor for TIM-1, also known as the Hepatitis A virus cellular receptor 1 (Havcr1).

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-4
Applications Flow Cytometry
Classification Monoclonal
Clone 21H12
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.