Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-4 Rabbit anti-Mouse, Clone: 3A1, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
| Antigen | TIM-4 |
|---|---|
| Clone | 3A1 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
TIM-4 Monoclonal antibody specifically detects TIM-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, NeutralizationSpecifications
| TIM-4 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
| Unconjugated | |
| RUO | |
| PBS | |
| 91937 | |
| IgG κ | |
| Protein A purified |
| 3A1 | |
| Monoclonal | |
| Rabbit | |
| Mouse | |
| FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
| This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with TIM-4:Ig fusion protein in complete Freund's adjuvant. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title