Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM17 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589308 Supplier Thermo Scientific Supplier No. PA589308

Rabbit Polyclonal Antibody

Immunogen sequence: MEEYAREPCP WRIVDDCGGA FTMGTIGGGI FQAIKGFRNS PVGVNHRLRG SLTAIKTRAP QLGGSFAVWG GLFSMIDCSM VQVRGKEDPW NSITSGALTG AILAARNGPV AMVGSAAMGG ILLALIEGAG ILLTRFASAQ FPNGPQFAED PSQLPSTQLP SSPFGDYRQY Q; Positive Samples: NCI-H460, HeLa, A-549, MCF-7, HT-29, Mouse kidnye, Mouse heart; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein

Translocation of nuclear encoded preproteins into the mitochondrial matrix requires the coordinated action of the translocases Tom and Tim, which are located in the outer mitochondrial membrane and the inner membrane, respectively. The mitochondrial preprotein translocases of the outer membrane (Tom) is a multi-subunit protein that contains at least eight proteins: four import receptor subunits (Tom70, Tom37, Tom22, and Tom20), three small proteins (Tom7, Tom6, and Tom5), and a structural component of the outer membrane channel (Tom40). The Tom machinery involves the import receptors, which initiate the binding of cytosolically synthesized preproteins to the outer membrane, and a general import pore (GIP), which promotes the translocation of various pre-proteins into the mitochondria. The TIM channel imports nuclear-encoded mitochondrial preproteins, and it involves three proteins, Tim17, Tim23 and Tim44, which are represented at equimolar ratios. Tim17 is expressed as two isoforms Tim17a and Tim17b, which differ only in their C termini sequences, and like Tim23, these proteins are ubiquitously expressed in fetal and adult tissues. Tim17 and Tim23 are integral membrane proteins that comprise the structural elements of the inner membrane channel by which the preproteins are transferred. The Tim44, on the other hand, is a largely hydrophilic protein that recruits the matrix located Hsp70 to the site where the preprotein emerges from the Tim channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM17
Applications Immunocytochemistry, Western Blot, Western Blot
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene TIMM17A
Gene Accession No. Q99595, Q9Z0V8
Gene Alias 17kDa, fb82c12, Inner membrane preprotein translocase Tim17a, Mimt17, Mitochondrial import inner membrane translocase subunit Tim17-A, Mitochondrial import inner membrane translocase subunit Tim17-A-like protein, mitochondrial inner membrane translocase, mTim17a, preprotein translocase, Tim17, TIM17A, TIMM17, TIMM17A, timm17a protein, translocase of inner mitochondrial membrane 17 homolog A, translocase of inner mitochondrial membrane 17 homolog A (yeast), translocase of inner mitochondrial membrane 17a, translocator of inner mitochondrial membrane 17 kDa, a, translocator of inner mitochondrial membrane 17a, translocator of inner mitochondrial membrane a, wu:fb82c12
Gene Symbols TIMM17A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-171 of human TIMM17A (NP_0063261)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10440, 21854
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.