Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMD2 Chimeric Recombinant Rabbit Monoclonal Antibody (RMT2-14)
GREENER_CHOICE

Catalog No. PIMA548167
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548167 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548167 Supplier Invitrogen™ Supplier No. MA548167
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody is specific for murine TIM-2, a transmembrane protein that is expressed on activated T cells and preferentially on differentiated Th2 cells. A blockade of TIM-2/TIM-2 ligand interaction results in T cell hyperproliferation and Th2 cytokine production. TIM-2 plays a critical role in the development of atopic disease and other Th2-biased immune responses.

TIM-2 recognizes T-cell immunoglobulin mucin 2, a transmembrane glycoprotein and member of the immunoglobulin superfamily, expressed by activated lymphocytes, developmentally in the 8-cell embryo and blastocyst, and preferentially up-regulated in Th2 cells and down-regulated in Th1 cells. Tim-2 acts as a negative regulator of Th2 cells, and is implicated to play a critical role in the development of atopic disease and other Th2-biased immune responses. Tim-2 also acts as a receptor for Semaphorin 4A, a transmembrane protein expressed on dendritic cells and B cells, which plays a role in the guidance of axonal migration during neuronal development, and has been shown to enhance T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMD2 Chimeric
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RMT2-14
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Timd2
Gene Accession No. Q8R183
Gene Alias AA816106; AU019358; AU019662; C78111; G9D3; T cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain-containing protein 2; T-cell immunoglobulin mucin receptor 2; T-cell membrane protein 2; Tim2; TIM-2; Timd2; TIMD-2
Gene Symbols Timd2
Host Species Rabbit
Immunogen This antibody was raised by immunising Sprague Dawley rats with TIM-2-Ig consisting of the extracellular domain (aa 1-230 of mouse TIM-2) and the Fc portion of mouse IgG2a, emulsified in complete Freund's adjuvant and then fusing lymph node cells from the immunized rats with P3U1 myeloma cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171284
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.