Learn More
Invitrogen™ TIMD2 Monoclonal Antibody (RMT2-25), FITC

Description
Rat anti Mouse TIM-2 antibody, clone RMT2-25 recognizes mouse Tim-2 (T-cell immunoglobulin mucin 2), a transmembrane glycoprotein and member of the immunoglobulin superfamily, expressed by activated lymphocytes, developmentally in the 8-cell embryo and blastocyst, and preferentially up-regulated in Th2 cells and down-regulated in Th1 cells.
Specifications
Specifications
| Antigen | TIMD2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RMT2-25 |
| Concentration | 100 μg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide |
| Gene | Timd2 |
| Gene Accession No. | Q8R183 |
| Gene Alias | AA816106; AU019358; AU019662; C78111; G9D3; T cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain-containing protein 2; T-cell immunoglobulin mucin receptor 2; T-cell membrane protein 2; Tim2; TIM-2; Timd2; TIMD-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.