Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMD2 Monoclonal Antibody (RMT2-25)
GREENER_CHOICE

Catalog No. PIMA528252
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528252 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528252 Supplier Invitrogen™ Supplier No. MA528252
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rat anti Mouse TIM-2 antibody, clone RMT2-25 recognizes mouse Tim-2 (T-cell immunoglobulin mucin 2), a transmembrane glycoprotein and member of the immunoglobulin superfamily, expressed by activated lymphocytes, developmentally in the 8-cell embryo and blastocyst, and preferentially up-regulated in Th2 cells and down-regulated in Th1 cells.

TIM-2 recognizes T-cell immunoglobulin mucin 2, a transmembrane glycoprotein and member of the immunoglobulin superfamily, expressed by activated lymphocytes, developmentally in the 8-cell embryo and blastocyst, and preferentially up-regulated in Th2 cells and down-regulated in Th1 cells. Tim-2 acts as a negative regulator of Th2 cells, and is implicated to play a critical role in the development of atopic disease and other Th2-biased immune responses. Tim-2 also acts as a receptor for Semaphorin 4A, a transmembrane protein expressed on dendritic cells and B cells, which plays a role in the guidance of axonal migration during neuronal development, and has been shown to enhance T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMD2
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RMT2-25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Timd2
Gene Accession No. Q8R183
Gene Alias AA816106; AU019358; AU019662; C78111; G9D3; T cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain containing 2; T-cell immunoglobulin and mucin domain-containing protein 2; T-cell immunoglobulin mucin receptor 2; T-cell membrane protein 2; Tim2; TIM-2; Timd2; TIMD-2
Gene Symbols Timd2
Host Species Rat
Immunogen Tim-2-Ig, consisting of amino acids 1-230 of mouse Tim-2, coupled with the Fc portion of mouse IgG2a.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171284
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.