Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMD4 Recombinant Rabbit Monoclonal Antibody (102)
GREENER_CHOICE

Catalog No. PIMA531239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531239 Supplier Thermo Scientific Supplier No. MA531239
Add to Cart
Add to Cart

TIMD4 Recombinant Rabbit Monoclonal Antibody (102)

TIMD4 Recombinant Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMD4
Applications ELISA
Classification Recombinant Monoclonal
Clone 102
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TIMD4
Gene Accession No. Q6U7R4
Gene Alias B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4
Gene Symbols TIMD4
Host Species Rabbit
Immunogen Recombinant Mouse TIM4/TIMD4 protein (Met1-Thr279).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 276891
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.