Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Timeless Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10040853
Description
Timeless Polyclonal specifically detects Timeless in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Timeless | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
FLJ12640, hTIMTIMELESS1, TIM1protein timeless homolog, timeless (Drosophila) homolog, timeless homolog (Drosophila), TIMFLJ20714 | |
Rabbit | |
139 kDa | |
0.1 mL | |
Primary | |
Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:250 | |
Q9UNS1 | |
TIMELESS | |
The immunogen recognized by this antibody maps to a region between residue 1170 and the C-terminus (residue 1208) of human Timeless using the numbering given in entry NP_003911.1 (GeneID 8914). | |
Affinity Purified | |
RUO | |
8914 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction