Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Timeless Antibody (SR1201), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $408.03
Specifications
| Antigen | Timeless |
|---|---|
| Clone | SR1201 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
Timeless Monoclonal antibody specifically detects Timeless in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Timeless | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ12640, hTIMTIMELESS1, TIM1protein timeless homolog, timeless (Drosophila) homolog, timeless homolog (Drosophila), TIMFLJ20714 | |
| A synthesized peptide derived from human Timeless (Uniprot #: Q9UNS1) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1201 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 8914 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title