Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMELESS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578270 Supplier Invitrogen™ Supplier No. PA578270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMELESS
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TIMELESS
Gene Accession No. Q9R1X4, Q9UNS1
Gene Alias C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown
Gene Symbols TIMELESS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human TIMELESS. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21853, 8914
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.