Learn More
Invitrogen™ TIMELESS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592175
Description
Immunogen sequence: EDLEEEENLP EEDSEEEEEG GSEAEQVQGS LVLSNENLGQ SLHQEGFSIP LLWLQNCLIR AADDREEDGC SQAVPLVPLT EENEEAMENE QFQQLLRKLG VRPPASGQET FWRIPAKLSP TQLRRAAASL SQPEEEQKLQ PELQPKVPGE QGSDEEHCKE HRAQALRALL LAHKKKAGLA SPEEEDAVGK EPLKAAPKKR QLLDSDEEQE EDEGRNRAPE LGAPGIQKKK RYQIEDDEDD; Positive Samples: BxPC-3; Cellular Location: Nucleus.
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifications
TIMELESS | |
Polyclonal | |
Unconjugated | |
TIMELESS | |
C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown | |
Rabbit | |
Affinity chromatography | |
RUO | |
21853, 8914 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9R1X4, Q9UNS1 | |
TIMELESS | |
Recombinant fusion protein containing a sequence corresponding to amino acids 969-1208 of human TIMELESS (NP_0039112). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.