Learn More
Invitrogen™ TIMELESS Recombinant Rabbit Monoclonal Antibody (24GB2665)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557778

Description
TIMELESS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifications
TIMELESS | |
Recombinant Monoclonal | |
Unconjugated | |
TIMELESS | |
C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown | |
Rabbit | |
Affinity chromatography | |
RUO | |
21853, 8914 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
24GB2665 | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9R1X4, Q9UNS1 | |
TIMELESS | |
A synthesized peptide derived from human Timeless (aa 1150-1208). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.