Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMM10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321445100UL
Description
TIMM10 Polyclonal antibody specifically detects TIMM10 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
TIMM10 | |
Polyclonal | |
Immunohistochemistry, Immunoprecipitation 1-5 uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
mitochondrial import inner membrane translocase subunit Tim10, TIM10TIM10Atranslocase of inner mitochondrial membrane 10 (yeast) homolog, translocase of inner mitochondrial membrane 10 homolog (yeast) | |
Produced in rabbits immunized with E. coli-derived Human TIMM10 fragment | |
100 μg | |
Endocrinology, Signal Transduction | |
26519 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction