Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559002 Supplier Invitrogen™ Supplier No. PA559002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPLRAQQLA AELEVEMMAD MYNRMTSACH RKCVPPHYKE AELSKGESVC LDRCVSKYLD IHERMGKKLT ELSMQDEELM KRVQQSSGPA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The mitochondrial protein encoded by this gene belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane, functioning as intermembrane space chaperones for the highly insoluble carrier proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Timm10
Gene Accession No. P62072
Gene Alias Mitochondrial import inner membrane translocase subunit Tim10; small zinc finger-like protein; Tim10; TIM10A; Tim13; TIMM10; Timm13a; translocase of inner mitochondrial membrane 10; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 10 homolog (yeast); translocase of inner mitochondrial membrane 13 homolog a
Gene Symbols Timm10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIMM10. Recombinant protein control fragment (Product #RP-97344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.