Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIMM10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110488
Description
Immunogen sequence: MDPLRAQQLA AELEVEMMAD MYNRMTSACH RKCVPPHYKE AELSKGESVC LDRCVSKYLD IHERMGKKLT ELSMQDEELM KRVQQSSGPA.
The mitochondrial protein encoded by this gene belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane, functioning as intermembrane space chaperones for the highly insoluble carrier proteins.
Specifications
TIMM10 | |
Polyclonal | |
Unconjugated | |
Timm10 | |
Mitochondrial import inner membrane translocase subunit Tim10; small zinc finger-like protein; Tim10; TIM10A; Tim13; TIMM10; Timm13a; translocase of inner mitochondrial membrane 10; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 10 homolog (yeast); translocase of inner mitochondrial membrane 13 homolog a | |
Rabbit | |
Affinity chromatography | |
RUO | |
26519, 30059, 64464 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62072, P62073, P62074 | |
Timm10 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human TIMM10 (NP_036588.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction