Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100352 Supplier Invitrogen™ Supplier No. PA5100352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIMM13.

This gene encodes a member of the evolutionarily conserved TIMM (translocase of inner mitochondrial membrane) family of proteins that function as chaperones in the import of proteins from the cytoplasm into the mitochondrial inner membrane. Proteins of this family play a role in collecting substrate proteins from the translocase of the outer mitochondrial membrane (TOM) complex and delivering them to either the sorting and assembly machinery in the outer mitochondrial membrane (SAM) complex or the TIMM22 complex in the inner mitochondrial membrane. The encoded protein and the translocase of mitochondrial inner membrane 8a protein form a 70 kDa complex in the intermembrane space.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM13
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIMM13
Gene Accession No. P62075, P62076, Q9Y5L4
Gene Alias D10Ertd378e; fd44h11; hypothetical protein LOC436738; mitochondrial import inner membrane translocase subunit Tim13; mitochondrial import inner membrane translocase subunit Tim13B; ppv1; TIM13; Tim13a; TIM13B; Tim9; Timm13; TIMM13A; TIMM13B; Timm9; translocase of inner mitochondrial membrane 13; translocase of inner mitochondrial membrane 13 homolog; translocase of inner mitochondrial membrane 13 homolog (yeast); translocase of inner mitochondrial membrane 9 homolog; wu:fd44h11; zgc:92895
Gene Symbols TIMM13
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIMM13(Accession Q9Y5L4), corresponding to amino acid residues D59-M95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252928, 26517, 30055
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.