Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMM17A Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297571PCP
Description
TIMM17A Polyclonal antibody specifically detects TIMM17A in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TIMM17A | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
Inner membrane preprotein translocase Tim17a, mitochondrial import inner membrane translocase subunit Tim17-A, mitochondrial inner membrane translocase, TIM17Apreprotein translocase, TIM17MIMT17, TIMM17, translocase of inner mitochondrial membrane 17 homolog A (yeast) | |
Produced in rabbits immunized with E. coli-derived Human TIMM17A fragment. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
10440 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction