Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM17B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106382 Supplier Invitrogen™ Supplier No. PA5106382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIMM17B.

This gene encodes a multipass transmembrane protein that forms an integral component of the mitochondrial translocase TIM23 complex. This complex facilitates the transport of mitochondrial proteins from the cytosol across the mitochondrial inner membrane and into the mitochondrion. There is a pseudogene for this gene on chromosome 12. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM17B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Timm17b
Gene Accession No. O60830, Q9Z0V7
Gene Alias 17kDa; DXS9822; inner mitochondrial membrane preprotein translocase; JM3; Mitochondrial import inner membrane translocase subunit Tim17-B; mTim17b; RGD1561744; Sfc3; TIM17B; TIMM17B; translocase of inner mitochondrial membrane 17 homolog B (yeast); translocase of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane 17 kDa, b; translocator of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane b
Gene Symbols Timm17b
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIMM17B(Accession O60830), corresponding to amino acid residues V42-S92.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10245, 21855, 317374
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.