Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIMM21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552710
Description
Immunogen sequence: LRAVQYTEKL HRSSAKRLLL PYIVLNKACL KTEPSLRCGL QYQKKTLRPR CILGVTQKTI WTQGPSPRKA KEDGSKQVSV HRSQRGGTAV PTSQKVKEAG RD Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 69%.
Involved in mitochondrial cytochrome c oxidase assembly; mitochondrial respiratory chain complex I assembly; and protein import into mitochondrial matrix. Part of TIM23 mitochondrial import inner membrane translocase complex.
Specifications
TIMM21 | |
Polyclonal | |
Unconjugated | |
TIMM21 | |
1700034H14Rik; 2700002I20Rik; C18orf55; HSPC154; Mitochondrial import inner membrane translocase subunit Tim21; Tim21; TIM21-like protein, mitochondrial; TIMM21; tranlocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29090 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVV7 | |
TIMM21 | |
Recombinant protein corresponding to Human TIMM21. Recombinant protein control fragment (Product #RP-89986). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction