Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562431
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562431 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562431 Supplier Invitrogen™ Supplier No. PA562431
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSSSSPSKIY GRALEKCRSH PEVIGVFGES VKGYGEVTRR GRRQHVRFTE YVKDGLKHTC VKFYIEGSEP GKQGTVYAQV KENPGSGEYD FRYIFVEIES YPRRTIIIED Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.

Involved in mitochondrial cytochrome c oxidase assembly; mitochondrial respiratory chain complex I assembly; and protein import into mitochondrial matrix. Part of TIM23 mitochondrial import inner membrane translocase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM21
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIMM21
Gene Accession No. Q9BVV7
Gene Alias 1700034H14Rik; 2700002I20Rik; C18orf55; HSPC154; Mitochondrial import inner membrane translocase subunit Tim21; Tim21; TIM21-like protein, mitochondrial; TIMM21; tranlocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21 homolog
Gene Symbols TIMM21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIMM21. Recombinant protein control fragment (Product #RP-103103).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29090
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.