Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560935 Supplier Invitrogen™ Supplier No. PA560935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIDTNVGFDP KDPYRTPTAK EVLKDMGQRG MSYAKNFAIV GAMFSCTECL IESYRGTSDW KNSVISGCIT GGAIGFRAGL KA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

TIM22 is a translocase localized in mitochondrial inner membrane. It is implicated in import of nuclear-encoded mitochondrial preproteins to the mitochondria. TIM22 is the essential core of protein insertion complex (TIM22 complex) that consists of TIM22, TIM54, TIM18, and TIM12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Timm22
Gene Accession No. Q9Y584
Gene Alias mitochondrial import inner membrane translocase subunit Tim22; preprotein translocase; putative membrane protein; Testis-expressed protein 4; testis-expressed sequence 4; TEX4; TIM22; TIMM22; translocase of inner mitochondrial membrane 22; translocase of inner mitochondrial membrane 22 homolog; translocase of inner mitochondrial membrane 22 homolog (yeast)
Gene Symbols Timm22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIMM22. Recombinant protein control fragment (Product #RP-102447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.