Learn More
Invitrogen™ TIMM22 Recombinant Rabbit Monoclonal Antibody (2H2R3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555576

Description
TIMM22 Recombinant Monoclonal Antibody for Western Blot, ELISA
TIM22 is a translocase localized in mitochondrial inner membrane. It is implicated in import of nuclear-encoded mitochondrial preproteins to the mitochondria. TIM22 is the essential core of protein insertion complex (TIM22 complex) that consists of TIM22, TIM54, TIM18, and TIM12.
Specifications
TIMM22 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CQ85, Q9JKW1, Q9Y584 | |
Timm22 | |
A synthetic peptide corresponding to a sequence within amino acids 20-120aa of human TIMM22. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
2H2R3 | |
Unconjugated | |
Timm22 | |
mitochondrial import inner membrane translocase subunit Tim22; preprotein translocase; putative membrane protein; Testis-expressed protein 4; testis-expressed sequence 4; TEX4; TIM22; TIMM22; translocase of inner mitochondrial membrane 22; translocase of inner mitochondrial membrane 22 homolog; translocase of inner mitochondrial membrane 22 homolog (yeast) | |
Rabbit | |
Protein A | |
RUO | |
29928, 56322, 79463 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.