Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIMM23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571877
Description
TIMM23 Polyclonal Antibody for Western Blot
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Specifications
TIMM23 | |
Polyclonal | |
Unconjugated | |
Timm23 | |
mitochondrial import inner membrane translocase subunit Tim23; TIM23; TIMM23; translocase of inner mitochondrial membrane 23; translocase of inner mitochondrial membrane 23 homolog; translocase of inner mitochondrial membrane 23 homolog (yeast); translocase of the inner mitochondrial membrane | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100287932 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O14925 | |
Timm23 | |
KLH conjugated synthetic peptide between 130-158 amino acids from the C-terminal region of human TIMM23. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction