Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598194 Supplier Invitrogen™ Supplier No. PA598194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIMM23 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM23
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Timm23
Gene Accession No. O14925, O35093, Q9WTQ8
Gene Alias mitochondrial import inner membrane translocase subunit Tim23; TIM23; TIMM23; translocase of inner mitochondrial membrane 23; translocase of inner mitochondrial membrane 23 homolog; translocase of inner mitochondrial membrane 23 homolog (yeast); translocase of the inner mitochondrial membrane
Gene Symbols Timm23
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial import inner membrane translocase subunit Tim23 protein (1-209AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100287932, 53600, 54312
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.