Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMM44 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TIMM44 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TIMM44 Polyclonal specifically detects TIMM44 in Human samples. It is validated for Western Blot.Specifications
TIMM44 | |
Polyclonal | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
MIMT44, mitochondrial import inner membrane translocase subunit TIM44, mitochondrial inner membrane translocase, TIM44DKFZp686H05241, translocase of inner mitochondrial membrane 44 homolog (yeast) | |
TIMM44 | |
IgG | |
This product is specific to Subunit or Isoform: TIM44. |
Western Blot | |
Unconjugated | |
RUO | |
O43615 | |
10469 | |
Synthetic peptides corresponding to TIMM44(translocase of inner mitochondrial membrane 44 homolog (yeast)) The peptide sequence was selected from the N terminal of TIMM44. Peptide sequence MAAAALRSGWCRCPRRCLGSGIQFLSSHNLPHGSTYQMRRPGGELPLSKS | |
Primary | |
51 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title