Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120549 Supplier Invitrogen™ Supplier No. PA5120549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, K562, mouse liver, rat kidney. Immunogen sequence: AKTAKQSAES VSKGGEKLGR TAAFRALSQG VESVKKEIDD SVLGQTGPYR RPQRLRKRTE FAGDKFKEEK VFEPNEEALG VVLHKDSKWY QQWKDFKENN VVFNRFFEMK MKYDESDNAF IRASRALTDK VTDLLGGLFS KTEMSEVLTE ILRVDPAFDK DRFLKQCEND IIPNVLEAMI SGELDILKDW CYEATYSQLA HPIQQAKALG LQFHSRILDI DNVDLAMGKM MEQGPVLIIT FQAQLVMVVR NPKGEVVEGD PDKVLRMLYV WALCRDQDEL NPYAAWRLLD ISASSTEQIL.

This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM44
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TIMM44
Gene Accession No. O35094, O35857, O43615
Gene Alias 0710005E20Rik; D8Ertd118e; Mimt44; Mitochondrial import inner membrane translocase subunit TIM44; Tim44; TIMM44; translocase of inner mitochondrial membrane 44; translocase of inner mitochondrial membrane 44 homolog; translocase of inner mitochondrial membrane 44 homolog (yeast); translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed
Gene Symbols TIMM44
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 153-452 of human TIMM44 (NP_006342.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10469, 21856, 29635
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.