Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518308
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518308 Supplier Invitrogen™ Supplier No. PA518308
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM50
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TIMM50
Gene Accession No. Q3ZCQ8
Gene Alias 2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357
Gene Symbols TIMM50
Host Species Goat
Immunogen Synthetic peptide sequence (CDVRTVLEHYALEDD) corresponding to the internal amino acids of TIMM50 (aa 293-306).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92609
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.