Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563305 Supplier Invitrogen™ Supplier No. PA563305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGWRFKKRPG IETLFQQLAP LYEIVIFTSE TGMTAFPLID SVDPHGFISY RLFRDATRYM DGHHVKDISC LNRDPARVVV VDC Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM50
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIMM50
Gene Accession No. Q3ZCQ8
Gene Alias 2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357
Gene Symbols TIMM50
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIM50. Recombinant protein control fragment (Product #RP-103257).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92609
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.