Learn More
Invitrogen™ TIMM50 Polyclonal Antibody

Description
Immunogen sequence: GNNPVDENGA KIPDEFDNDP ILVQQLRRTY KYFKDYRQMI IEPTSPCLLP DPLQEPYYQP PYTLVLELTG VLLHPEWSLA TGWRFKKRPG IETLFQQLAP LYEIVIFTSE TGMTAFPLID SVDPHGFISY RLFRDATRYM DGHHVKDISC LNRDPARVVV VDCKKEAFRL QPYNGVALRP WDGNSDDRVL LDLSAFLKTI ALNGVEDVRT VLEHYALEDD PLAAFKQRQS RLEQEEQQRL AELSKSNKQN LFLGSLTSRL WPRSKQP.
Specifications
Specifications
| Antigen | TIMM50 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.83 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TIMM50 |
| Gene Accession No. | Q3ZCQ8, Q9D880 |
| Gene Alias | 2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357 |
| Gene Symbols | TIMM50 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.