Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIMM50 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546303
Description
Peptide sequence: EDDPLAAFKQ RQSRLEQEEQ QRLAELSKSN KQNLFLGSLT SRLWPRSKQP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 75%.
TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
Specifications
TIMM50 | |
Polyclonal | |
Unconjugated | |
TIMM50 | |
2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357 | |
Rabbit | |
Affinity chromatography | |
RUO | |
92609 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3ZCQ8 | |
TIMM50 | |
Synthetic peptide directed towards the C-terminal of human TIMM50 (aa 304-353). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction