Learn More
TIMM8A Mouse anti-Human, Clone: OTI5C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809738
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mitochondrial intermembrane chaperone that participates in the import and insertion of some multi-pass transmembrane proteins into the mitochondrial inner membrane. Also required for the transfer of beta-barrel precursors from the TOM complex to the sorting and assembly machinery (SAM complex) of the outer membrane. Acts as a chaperone-like protein that protects the hydrophobic precursors from aggregation and guide them through the mitochondrial intermembrane space. The TIMM8-TIMM13 complex mediates the import of proteins such as TIMM23, SLC25A12/ARALAR1 and SLC25A13/ARALAR2, while the predomit TIMM9-TIMM10 70 kDa complex mediates the import of much more proteins. Probably necessary for normal neurologic development.Specifications
| TIMM8A | |
| Monoclonal | |
| Unconjugated | |
| TIMM8A | |
| DDP, DDP1, DFN1, MTS, TIM8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1678 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O60220 | |
| TIMM8A | |
| Full length human recombit protein of human TIMM8A produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.