Learn More
Invitrogen™ TIMM8A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597251
Description
Immunogen sequence: MDSSSSSSAA GLGAVDPQLQ HFIEVETQKQ RFQQLVHQMT ELCWEKCMDK PGPKLDSRAE ACFVNCVERF IDTSQFILNR LEQTQKSKPV FSESLSD; Positive Samples: U-251MG, A-549, LO2, 293T, MCF-7, HeLa, Mouse liver, Mouse brain; Cellular Location: Intermembrane side, Mitochondrion inner membrane, Peripheral membrane protein.
Mitochondrial intermembrane chaperone that participates in the import and insertion of some multi-pass transmembrane proteins into the mitochondrial inner membrane. Also required for the transfer of beta-barrel precursors from the TOM complex to the sorting and assembly machinery (SAM complex) of the outer membrane. Acts as a chaperone-like protein that protects the hydrophobic precursors from aggregation and guide them through the mitochondrial intermembrane space. The TIMM8-TIMM13 complex mediates the import of proteins such as TIMM23, SLC25A12/ARALAR1 and SLC25A13/ARALAR2, while the predominant TIMM9-TIMM10 70 kDa complex mediates the import of much more proteins. Probably necessary for normal neurologic development.
Specifications
TIMM8A | |
Polyclonal | |
Unconjugated | |
TIMM8A | |
DDP; Ddp1; deafness dystonia protein 1; Deafness dystonia protein 1 homolog; deafness/dystonia peptide; DFN1; DXHXS1274E; Fci-12; Mitochondrial import inner membrane translocase subunit Tim8 A; MTS; TIM8; Tim8a; Timm8a; Timm8a1; translocase of inner mitochondrial membrane 8 homolog a; translocase of inner mitochondrial membrane 8 homolog A (yeast); translocase of inner mitochondrial membrane 8 homolog a1; translocase of inner mitochondrial membrane 8 homolog A1 (yeast); translocase of inner mitochondrial membrane 8A1; X-linked deafness dystonia protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1678, 30058, 84383 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60220, Q9WVA1, Q9WVA2 | |
TIMM8A, Timm8a1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-97 of human TIMM8A (NP_0040761). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.