Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM8A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1653 Supplier Bioss Supplier No. BS11769R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mitochondrial intermembrane chaperone that participates in the import and insertion of some multi-pass transmembrane proteins into the mitochondrial inner membrane. Also required for the transfer of beta-barrel precursors from the TOM complex to the sorting and assembly machinery (SAM complex) of the outer membrane. Acts as a chaperone-like protein that protects the hydrophobic precursors from aggregation and guide them through the mitochondrial intermembrane space. The TIMM8-TIMM13 complex mediates the import of proteins such as TIMM23, SLC25A12/ARALAR1 and SLC25A13/ARALAR2, while the predominant TIMM9-TIMM10 70 kDa complex mediates the import of much more proteins. Probably necessary for normal neurologic development.

Specifications

Antigen TIMM8A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TIMM8A
Gene Accession No. O60220
Gene Alias DDP; DDP1; Deafness dystonia protein 1; Deafness dystonia protein 1 homolog; deafness/dystonia peptide; DFN1; DXHXS1274E; Fci-12; Mitochondrial import inner membrane translocase subunit Tim8 A; MTS; TIM8; TIM8A; TIMM8A; Timm8a1; translocase of inner mitochondrial membrane 8 homolog a; translocase of inner mitochondrial membrane 8 homolog A (yeast); translocase of inner mitochondrial membrane 8 homolog a1; translocase of inner mitochondrial membrane 8 homolog A1 (yeast); translocase of inner mitochondrial membrane 8A1; X-linked deafness dystonia protein
Gene Symbols TIMM8A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TIMM8A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1678
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.