Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TIMM8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110490
Description
Immunogen sequence: MAELGEADEA ELQRLVAAEQ QKAQFTAQVH HFMELCWDKC VEKPGNRLDS RTENCLSSCV DRFIDTTLAI TSRFAQIVQK GGQ.
This gene encodes a member of a well-conserved family of proteins with similarity to yeast Tim mitochondrial import proteins. This gene is encoded by a nuclear gene and is transported into the intermembrane space of the mitochondrion. When formed into complexes, these proteins guide membrane-spanning proteins across the mitochondrial intermembrane space before they are added into the mitochondrial inner membrane. This gene is adjacent to succinate dehydrogenase, subunit D (SDHD), in which mutations have been found in affected members of families with hereditary paraganglioma.
Specifications
TIMM8B | |
Polyclonal | |
Unconjugated | |
TIMM8B | |
Ddp2; DDPL; DDP-like protein; Deafness dystonia protein 2; deafness dystonia protein 2 homolog; Mitochondrial import inner membrane translocase subunit Tim8 B; small zinc finger-like protein DDP2; TIM8B; TIMM8B; translocase of inner mitochondrial membrane 8 homolog b; translocase of inner mitochondrial membrane 8 homolog b (yeast); translocase of inner mitochondrial membrane 8B | |
Rabbit | |
Affinity chromatography | |
RUO | |
26521, 30057, 64372 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.59 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P62077, P62078, Q9Y5J9 | |
TIMM8B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 16-98 of human TIMM8B (NP_0365911). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction