Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559466 Supplier Invitrogen™ Supplier No. PA559466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGEADEAELQ RLVAAEQQKA QFTAQVHHFM ELCWDKCVEK PGNRLDSRTE NCLSSCVDRF IDTTLAITSR FAQIVQKGG Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a member of a well-conserved family of proteins with similarity to yeast Tim mitochondrial import proteins. This gene is encoded by a nuclear gene and is transported into the intermembrane space of the mitochondrion. When formed into complexes, these proteins guide membrane-spanning proteins across the mitochondrial intermembrane space before they are added into the mitochondrial inner membrane. This gene is adjacent to succinate dehydrogenase, subunit D (SDHD), in which mutations have been found in affected members of families with hereditary paraganglioma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM8B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIMM8B
Gene Accession No. Q9Y5J9
Gene Alias Ddp2; DDPL; DDP-like protein; Deafness dystonia protein 2; deafness dystonia protein 2 homolog; Mitochondrial import inner membrane translocase subunit Tim8 B; small zinc finger-like protein DDP2; TIM8B; TIMM8B; translocase of inner mitochondrial membrane 8 homolog b; translocase of inner mitochondrial membrane 8 homolog b (yeast); translocase of inner mitochondrial membrane 8B
Gene Symbols TIMM8B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIMM8B. Recombinant protein control fragment (Product #RP-97380).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.