Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106441 Supplier Invitrogen™ Supplier No. PA5106441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIMM9.

TIMM9 belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIMM9
Gene Accession No. Q9WV97, Q9WV98, Q9Y5J7
Gene Alias 2810011L15Rik; Mitochondrial import inner membrane translocase subunit Tim9; Tim10a; TIM9; TIM9A; Timm10; Timm9; Timm9a; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 9; translocase of inner mitochondrial membrane 9 homolog; translocase of inner mitochondrial membrane 9 homolog (yeast)
Gene Symbols TIMM9
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIMM9(Accession Q9Y5J7), corresponding to amino acid residues L53-R89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171139, 26520, 30056
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.