Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMMDC1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326071B
Description
TIMMDC1 Polyclonal antibody specifically detects TIMMDC1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
TIMMDC1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
chromosome 3 open reading frame 1, FLJ22597, M5-14 protein, Protein M5-14, transmembrane protein C3orf1 | |
Produced in rabbits immunized with E. coli-derived Human TIMMDC1 fragment | |
0.1 mL | |
Cell Biology | |
51300 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Biotin | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction