Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Antibody (OTI1A6), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274521APC
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.Specifications
| TIMP-2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| TIMP2 | |
| Full length recombinant protein of human TIMP-2 (NP_003246) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI1A6 | |
| Flow Cytometry | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers | |
| 7077.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction