Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Antibody (OTI1A6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201573
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIMP-2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
TIMP2 | |
Full length human recombinant protein of human TIMP2(NP_003246) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1A6 | |
Unconjugated | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
Mouse | |
21.7 kDa | |
0.1 mL | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers | |
7077 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction