Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Antibody (TIMP2/2044), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308714AF594
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
TIMP-2 | |
Monoclonal | |
Alexa Fluor 594 | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
Recombinant human TIMP-2 protein fragment (aa 48-220) (exact sequence is proprietary) (Uniprot: P16035) | |
100 μg | |
Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
7077 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin), Protein Array | |
TIMP2/2044 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction