Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-2 Antibody (MM0035-2C13), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11061001AF488
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIMP-2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TIMP2 | |
Human recombinant TIMP-2 | |
0.1 mL | |
Primary | |
No cross-reactivity to human TIMP-1 is noted | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0035-2C13 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers | |
7077 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction