Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP-2 Rabbit anti-Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish, Clone: TIMP2/2488R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen TIMP-2
Clone TIMP2/2488R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122922
SDP
View Documents
Novus Biologicals
NBP30772420UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

TIMP-2 Monoclonal specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

TIMP-2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers
10 mM PBS with 0.05% BSA
7077
IgG
Protein A or G purified
TIMP2/2488R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2
A synthetic peptide from the N-terminus of human TIMP-2 protein (aa56-66) (DSGNDIYGNPI)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.