Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-3 Antibody (MM0036-7D3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB110610020.025MG
Description
TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIMP-3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
TIMP3 (MM0036-7D3) | |
Human | |
Purified |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
MM0036-7D3 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
TIMP3 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant human tissue inhibitor of metalloproteinases-3 (TIMP-3). | |
0.025 mg | |
Angiogenesis, Apoptosis, Cancer, Hypoxia, Vision | |
7078 | |
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction