Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32143425UL
Description
TIMP-3 Polyclonal antibody specifically detects TIMP-3 in Human samples. It is validated for ImmunoprecipitationSpecifications
TIMP-3 | |
Polyclonal | |
Immunoprecipitation 1-5 uL/mg of lysate | |
HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
Produced in rabbits immunized with E. coli-derived Human TIMP-3 fragment | |
25 μg | |
Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
7078 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunoprecipitation | |
Unconjugated | |
PBS, pH 7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction