Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2294780.2MG
Description
TIMP-3 Polyclonal specifically detects TIMP-3 in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TIMP-3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
Rabbit | |
30 kDa | |
0.2 mg | |
Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
7078 | |
Human, Bovine, Canine, Equine, Sheep | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.2mg/mL | |
Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/Immunofluorescence 1-2ug/mlimmunohistochemistry 1-2ug/ml, Immunohistochemistry-Paraffin | |
P35625 | |
TIMP3 | |
A portion of amino acids 175-211 of human TIMP3 was used as immunogen for this antibody. | |
Protein A purified | |
RUO | |
Primary | |
TIMP3 (tissue inhibitor of metalloproteinases 3), along with family members TIMP1, TIMP2, and TIMP4, are inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix (ECM). An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors. TIMP s inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP-3 may be involved in regulating trophoblastic invasion of the uterus as well as in regulating remodeling of the extracellular matrix during the folding of epithelia, and in the formation, branching and expansion of epithelial tubes. | |
Store at 4C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction