Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP-4 Antibody (MM0037-9F3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11061003AF405
Description
TIMP-4 Monoclonal specifically detects TIMP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIMP-4 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TIMP4 | |
Human recombinant TIMP-4 | |
0.1 mL | |
Primary | |
No cross-reactivity to human TIMP-1, 2 and 3 is noted | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0037-9F3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cellular Markers, Hypoxia | |
7079 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction