Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMP1 Monoclonal Antibody (C8), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551017
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551017 200 μL
FEMA551018 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551017 Supplier Invitrogen™ Supplier No. MA551017
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP1
Applications Western Blot
Classification Monoclonal
Clone C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene TIMP1
Gene Accession No. P12032, P30120
Gene Alias CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1
Gene Symbols TIMP1
Host Species Mouse
Immunogen Recombinant Rat TIMP1 protein, Cys24-Ala217 (Accession #P30120).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116510, 21857
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.