Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMP2 Monoclonal Antibody (OTI1B11), TrueMAB™, OriGene
SDP

Catalog No. 502540539
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0539 30 μL
50-167-4815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0539 Supplier OriGene Supplier No. TA504044S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIMP2 Monoclonal antibody specifically detects TIMP2 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TIMP2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B11
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TIMP2
Gene Accession No. P16035, P25785, P30121, Q9TTY1
Gene Alias Collagenase inhibitor; CSC-21K; D11Bwg1104e; DDC8; etID32613.12; fa96h09; Metalloproteinase inhibitor 2; metalloproteinase inhibitor 2a; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; metalloproteinase-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP metallopeptidase inhibitor 2a; TIMP2; Timp-2; TIMP-2 protein; timp2a; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase 2 precursor; tissue inhibitor of metalloproteinase 2a; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinase-2 precursor; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2; wu:fa96h09; zgc:136726
Host Species Mouse
Immunogen Full length human recombinant protein of human TIMP2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21858, 29543, 403633, 7077
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.