Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMP3 Polyclonal Antibody

Catalog No. PIPA532621
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532621 500 μL
01-674-740 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA532621 Supplier Invitrogen™ Supplier No. PA532621

Rabbit Polyclonal Antibody

This antibody is predicted to react with rabbit and baboon based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or breast carcinoma.

Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene TIMP3
Gene Accession No. P35625
Gene Alias HSMRK222; K222; K222TA2; Metalloproteinase inhibitor 3; MIG-5 protein; protein MIG-5; SFD; Sun; TIMP 3; TIMP metallopeptidase inhibitor 3; TIMP metallopeptidase inhibitor 3 (Sorsby fundus dystrophy, pseudoinflammatory); TIMP metallopeptidase inhibitor 3 L homeolog; timp3; TIMP-3; TIMP-3 protein; timp3.L; timp3-A; tissue inhibitor metalloproteinase-3; tissue inhibitor of metalloproteinase 3; tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, pseudoinflammatory); tissue inhibitor of metalloproteinase-3; tissue inhibitor of metalloproteinase-3 precursor; tissue inhibitor of metalloproteinases 3; tissue inhibitor of metalloproteinases-3; XELAEV_18002484mg
Gene Symbols TIMP3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminus of human TIMP-3.
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7078
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.