Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578423 Supplier Invitrogen™ Supplier No. PA578423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse kidney Predicted Reactivity: Human (100%), Rat (97%), Xenopus laevis (83%), Dog (100%), Rabbit (97%), Chicken (82%), Sheep (100%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TIMP3
Gene Accession No. P35625, P39876, P48032
Gene Alias HSMRK222; K222; K222TA2; Metalloproteinase inhibitor 3; MIG-5 protein; protein MIG-5; SFD; Sun; TIMP 3; TIMP metallopeptidase inhibitor 3; TIMP metallopeptidase inhibitor 3 (Sorsby fundus dystrophy, pseudoinflammatory); TIMP metallopeptidase inhibitor 3 L homeolog; timp3; TIMP-3; TIMP-3 protein; timp3.L; timp3-A; tissue inhibitor metalloproteinase-3; tissue inhibitor of metalloproteinase 3; tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, pseudoinflammatory); tissue inhibitor of metalloproteinase-3; tissue inhibitor of metalloproteinase-3 precursor; tissue inhibitor of metalloproteinases 3; tissue inhibitor of metalloproteinases-3; XELAEV_18002484mg
Gene Symbols TIMP3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TIMP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21859, 25358, 7078
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.